Oncomine™ Tumor Mutation Load Assay
|
Variant type |
Oncomine™ Gene class |
Oncomine™ Variant class |
Annotation criteria |
|---|---|---|---|
|
Loss-of-function truncating de novo mutation |
Loss-of-Function |
Truncating |
|
|
Missense hotspot mutation |
Gain-of-Function Loss-of-Function |
Hotspot |
|
|
In-frame hotspot mutation |
Gain-of-Function Loss-of-Function |
Hotspot |
|
|
Splice site hotspot mutation |
Gain-of-Function Loss-of-Function |
Hotspot |
|
|
Intronic hotspot mutation |
Gain-of-Function Loss-of-Function |
Hotspot |
|
|
MNV hotspot mutation |
Gain-of-Function Loss-of-Function |
Hotspot |
|
|
CEBPA‑bZIP[1] |
Loss-of-Function |
CEBPA‑bZIP |
|
|
CEBPA‑N‑terminal [1] |
Loss-of-Function |
CEBPA‑N‑terminal |
|
|
EGFR exon 19 deletion [1] |
Gain-of-Function |
EGFRExon19Deletion |
|
|
EGFR exon 20 insertion [1] |
Gain-of-Function |
EGFRExon20Insertion |
|
|
ERBB2 exon 20 insertion [1] |
Gain-of-Function |
ERBB2Exon20Insertion |
|
|
JAK2 exon 12 indel [1] |
Gain-of-Function |
JAK2Exon12Indel |
|
|
KIT exon 11 deletion [1] |
Gain-of-Function |
KITExon11Deletion |
|
|
KIT exon 11 insertion [1] |
Gain-of-Function |
KITExon11Insertion |
|
|
MET exon 14 skipping [1] |
Gain-of-Function |
METExon14Skipping |
|
|
PDGFRA exon 18 deletion [1] |
Gain-of-Function |
PDGFRAExon18Deletion |
|