Oncomine™ Lung Cell‑Free Total Nucleic Acid Research Assay
|
Variant type |
Oncomine™ Gene class |
Oncomine™ Variant class |
Annotation criteria |
|---|---|---|---|
|
Copy number amplification |
Gain-of-Function |
Amplification |
|
|
Gene fusion |
Gain-of-Function |
Fusion |
|
|
Non-targeted fusion |
Gain-of-Function |
NonTargetedFusion |
|
|
RNA exon variant |
Gain-of-Function |
RNAExonVariant |
|
|
Missense hotspot mutation |
Gain-of-Function Loss-of-Function |
Hotspot |
|
|
In-frame hotspot mutation |
Gain-of-Function Loss-of-Function |
Hotspot |
|
|
Splice site hotspot mutation |
Gain-of-Function Loss-of-Function |
Hotspot |
|
|
Intronic hotspot mutation |
Gain-of-Function Loss-of-Function |
Hotspot |
|
|
Synonymous hotspot mutation |
Gain-of-Function Loss-of-Function |
Hotspot |
|
|
Gain-of-function truncating hotspot mutation |
Gain-of-Function |
Hotspot |
|
|
Loss-of-function truncating hotspot mutation |
Loss-of-Function |
Truncating |
|
|
MNV hotspot mutation |
Gain-of-Function Loss-of-Function |
Hotspot |
|
|
EGFR exon 19 deletion[1] |
Gain-of-Function |
EGFRExon19Deletion |
|
|
EGFR exon 20 insertion [1] |
Gain-of-Function |
EGFRExon20Insertion |
|
|
ERBB2 exon 20 insertion [1] |
Gain-of-Function |
ERBB2Exon20Insertion |
|
|
MET exon 14 skipping [1] |
Gain-of-Function |
METExon14Skipping |
|
| Value | ||
|---|---|---|
| Function, transcript, and coding syntax occur in predefined truncating hotspot list |