Oncomine BRCA Research Assay
Variant type
Oncomine Gene class
Oncomine Variant class
Annotation criteria
Copy number deletion
Loss-of-Function
Deletion
  • SVTYPE = "CNV"
  • FILTER = "LOSS"
  • SUBTYPE = "GeneCNV"
Copy number exon deletion
Loss-of-Function
ExonDeletion
  • SVTYPE = "CNV"
  • FILTER = "LOSS"
  • SUBTYPE = "BigDel"
Copy number exon duplication
Loss-of-Function
ExonDuplication
  • SVTYPE = "CNV"
  • FILTER = "GAIN"
  • SUBTYPE = "BigDup"
Long deletion
Loss-of-Function
Truncating
  • Positive mutation call
  • SVTYPE = "LongDel"
Loss-of-function truncating de novo mutation
Loss-of-Function
Truncating
  • Positive mutation call
  • Functional impact is frameshift block substitution, frameshift deletion, frameshift insertion, or nonsense
  • Occurs in a designated loss-of-function gene
Missense hotspot mutation
Loss-of-Function
Hotspot
  • Positive mutation call
  • Functional impact is missense
  • Transcript and codon position occur in predefined missense hotspot list
Splice site hotspot mutation
Loss-of-Function
Hotspot
  • Positive mutation call
  • Transcript, location, and exon occur in predefined splice site hotspot list