Oncomine™ Colon cfDNA Assay
Variant type
Oncomine™ Gene class
Oncomine™ Variant class
Annotation criteria
Missense hotspot mutation
Gain-of-Function
Loss-of-Function
Hotspot
  • Positive mutation call
  • Functional impact is missense
  • Transcript and codon position occur in predefined missense hotspot list
In-frame hotspot mutation
Gain-of-Function
Loss-of-Function
Hotspot
  • Positive mutation call
  • Function, transcript, and coding syntax occur in predefined in-frame hotspot list
Loss-of-function truncating hotspot mutation
Loss-of-Function
Truncating
  • Positive mutation call
  • Function, transcript, and coding syntax occur in predefined truncating hotspot list
  • Occurs in a loss-of-function gene
MNV hotspot mutation
Gain-of-Function
Loss-of-Function
Hotspot
  • Positive mutation call
  • Transcript and coding syntax occur in MNV hotspot list
ERBB2 exon 20 insertion[1]
Gain-of-Function
ERBB2Exon20Insertion
  • Positive mutation call
  • Functional impact is nonframeshift insertion, nonframeshift block substitution
  • Insertion impacts codons 770–783 of ERBB2[2]
1 For more information, see .
2 In some cases the assay design may not cover the entire codon range.