Oncomine™ Myeloid Research Assays
|
Variant type |
Oncomine™ Gene class |
Oncomine™ Variant class |
Annotation criteria |
|---|---|---|---|
|
Gene fusion |
Gain-of-Function |
Fusion |
|
|
Non-targeted fusion |
Gain-of-Function |
NonTargetedFusion |
|
|
RNA exon variant |
Gain-of-Function |
RNAExonVariant |
|
|
Loss-of-function truncating de novo mutation |
Loss-of-Function |
Truncating |
|
|
Missense hotspot mutation |
Gain-of-Function Loss-of-Function |
Hotspot |
|
|
In-frame hotspot mutation |
Gain-of-Function Loss-of-Function |
Hotspot |
|
|
Intronic hotspot mutation |
Gain-of-Function Loss-of-Function |
Hotspot |
|
|
Gain-of-function truncating hotspot mutation |
Gain-of-Function |
Hotspot |
|
|
Loss-of-function truncating hotspot mutation |
Loss-of-Function |
Truncating |
|
|
MNV hotspot mutation |
Gain-of-Function Loss-of-Function |
Hotspot |
|
|
CEBPA‑bZIP[1] |
Loss-of-Function |
CEBPA‑bZIP |
|
|
CEBPA‑N‑terminal [1] |
Loss-of-Function |
CEBPA‑N‑terminal |
|
|
JAK2 exon 12 indel [1] |
Gain-of-Function |
JAK2Exon12Indel |
|
|
KIT exon 11 deletion [1] |
Gain-of-Function |
KITExon11Deletion |
|
|
KIT exon 11 insertion [1] |
Gain-of-Function |
KITExon11Insertion |
|
|
FLT3 internal tandem duplication |
Gain-of-Function |
FLT3ITD |
|