Oncomine Breast cfDNA Assay
Variant type
Oncomine gene class
Oncomine variant class
Annotation criteria
Missense hotspot mutation
Gain-of-Function
Loss-of-Function
Hotspot
  • Positive mutation call
  • Functional impact is missense
  • Transcript and codon position occur in predefined missense hotspot list
In-frame hotspot mutation
Gain-of-Function
Loss-of-Function
Hotspot
  • Positive mutation call
  • Function, transcript, and coding syntax occur in predefined in-frame hotspot list