View Oncomine™ BRCA analysis results
Some Oncomine™ assays used with Ion Reporter™ Software include BRCA1 and BRCA2 amplicons that enable detection and visualization of whole exon and multiple exon deletion in BRCA1 and BRCA2 genes in somatic and germline samples with high sensitivity.
  1. In the Analyses tab, click Overview.
  2. Search, sort, or filter the Analyses table to find the sample result of interest.
  3. In the Analysis column, click the hyperlink for the analysis of interest to open Analysis Results screen for the BRCA analysis.
  4. The following data for the BRCA results is available in the Analysis Results screen.
    • A summary of called variants and their genotypic and functional properties. Variants that are listed include SNVs, INDELs, long exon deletions, duplications, and whole gene deletions and duplications.
    • Metrics and information at the top of the screen.
      Item
      Description
      Analysis Name
      The name of the analysis.
      Cancer Type
      The type of cancer, as defined by the sample attribute in Torrent Suite™ Software, if sample data are transferred by Torrent Suite™ Software.
      MAPD
      The MAPD (Median of the Absolute values of all Pairwise Differences) metric is an estimate of coverage variability between adjacent amplicons. The default threshold is 0.5. As a result, sample results with a MAPD above this value should be viewed with lower confidence.
      BRCA CNV QC
      A quality control score that is based on the MAPD threshold, and quality scores for the percent of non-zero amplicons, total number of reads, and the number of calls. BRCA CNV QC is PASSED or FAILED. If the BRCA CNV QC fails, a reason is provided.
      Sample ID
    • Click Summary to view a summary of the called variants. Select a classification from the dropdown list to assign a classification to a variant. The following variant types are available for a BRCA analysis.
      Type
      CNV Subtype
      Description
      CNV
      BigDel
      Deletion of at least one exon
      BigDup
      Duplication of at least one exon
      GeneCNV
      Whole BRCA1/BRCA2 gene deletion or duplication
      NOCALL
      Read count differs from baseline by non-integer amount; evidence for a BigDel or BigDup call is weak
      REF
      Read count matches reference baseline
      LongDel
      —
      A specific 40 bp deletion in BRCA1 (c.1176_1214del)
      SNV
      —
      Single nucleotide substitution
      MNP
      —
      Multiple nucleotide polymorphism at adjacent nucleotide positions
      INDEL
      —
      Single or multiple nucleotide insertion or deletion
    • Click Functional to view other functional annotations and use the annotations to classify, sort, and filter variants.
    • Click Pharmacogenomics to view the ClinVar column. Click the link in the ClinVar column for a selected variant to open an NCBI ClinVar website where information about the ClinVar variant annotation is available.
  5. Click Visualize to view a BRCA Report that shows read counts of BRCA1 and BRCA2 exons that are normalized to the Oncomine BRCA DNA Baseline. For more information, see Visualize the BRCA report.