Customize the Analysis Results table
The Analysis Results table includes details that describe the results of the analysis, samples, analysis workflows, and software versions that were used to create the analyses in Ion Reporter™ Software.
When you open an analysis, the information that is shown in the Analysis Results table depends on which workflow and annotation set is applied when the analysis is launched. You can customize the table to reflect the information that is most relevant to your research. To do so, create a table preference, or drag and drop the columns to change the order in which they appear.