Restore default settings for the Analysis Results table
You can restore default column headings in the Analysis Results table Ion Reporter™ Software.
  1. In the Analyses tab, click Overview.
  2. Click the link for an analysis of interest to open the Analysis Results table.
  3. Click PreferencesRestore Defaults.
The default table is restored.