View tumor mutational burden analysis results
You can view the results and calculated tumor mutational burden (TMB mutations/Mb) when you open analysis results that used a workflow with tumor mutational burden enabled.
Note: Microsoft™ Excel™, or other spreadsheet tool, is required for viewing VCF, CSV, and TSV files.
- Click the Analyses tab.
- In the Overview screen, use the Workflow filter to limit the list of analyses to limit the list of analyses that use an Oncomine™ Tumor Mutation Load workflow.
-
Click a row in the Analyses table to view the Details of the analysis.
In the Details pane you can view Workflow Details, and access the Actions list.
-
Click the link for the name of the Analysis to open the Analysis Results screen.
The Analysis Results screen opens to the Oncomine tab that displays Oncomine™ annotated variants that are relevant to cancer with Oncomine™ Gene Class and Oncomine™ Variant Class information.
Information for tumor mutational burden that is displayed at the top of the Analysis Results screen is as follows.
Item
Description
Tumor Mutational Burden (Mutations/Mb)result
The value of the result for the tumor mutational burden calculation.
For more information, see Example: Tumor Mutational Burden calculation.
TMB classification
A score of High, Intermediate, or Low.
Thresholds to determine the TMB classification are set with the TMB-Low Threshold and TMB-High Threshold parameters. For more information, see Annotation parameters.
A classification status of Undefined indicates that either the TMB classification thresholds settings are not set in the analysis workflow or were set incorrectly.
Note: Threshold values are not provided in the predefined analysis workflow. The analysis workflow always reports Undefined as the TMB classification status unless a user sets the thresholds in the parameter settings. -
In the Analysis Results table, sort or filter the data using the Oncomine™-specific annotations.
-
To download a results file.
The software generates a ZIP file with 4 folders: RESULTS, QC, Variants, and Workflow_Settings.
|
Folder |
Contents of folder |
|---|---|
|
RESULTS |
Note: The Variant Details TSV file can be downloaded separately. For more information see Visualize tumor mutational burden analysis results.. If the Tumor Mutational Burden parameter is enabled, or an analysis workflow for the Tumor Mutation Load Assay is used, a Results folder is generated that contains:
|
|
QC |
Contains a PDF of the QC report, and a folder containing coverage statistics files. |
|
Variants |
|
|
Workflow_Settings |
Contains folders with:
|