View the tumor mutational burden statistics
The tumor mutational burden value is calculated automatically when you launch select Oncomine™ analysis workflows, such as analysis workflows for Oncomine™ Comprehensive Assay Plus, DNA or Oncomine™ Tumor Mutation Load Assay. You can download the tumor mutational burden statistics to view more details of the analysis, such as the TMB algorithm version that is used, and information about how the tumor mutational burden is calculated.
  1. Download, then extract a results file to your hard drive.
    For more information, see Download analysis files.
  2. In the RESULTS folder, open the statistic.txt file in a compatible text editor. The tumor mutational burden and the data that are used to calculate its value are listed.
    In this example, the tumor mutational burden value is calculated from the values for Total Exonic Bases with Sufficient Depth of Base Coverage, and Total Somatic Filtered Variants Count (numerator for TMB calculation). TMB Standardization is off by default, that is, 0.0TMB Standardization='Not Applied'. Observed TMB score was set to 25, which is below the calibration threshold, so that the germline calibration was also not applied. That is Germline Calibration='Not Applied'.
    IR Sample Name='JH_TMB_colon1_4_N.bam.JH_TEST_BAM'IR
    Analysis Name='JH_TMB_colon1_4_N.bam.JH_TEST_BAM_1564544352260'Released with Ion Reporter 'ir512'.
    TMB Algorithm Version='V4.0'Minimum Read Depth of Base Coverage required=60.0
    Maximum Deamination Score allowed=100.0
    Minimum Variant Allele Frequency required=0.05
    Deamination=16 (QC: PASS; observed (16) < threshold (100.0))
    Total Bases (sequenced bases in the panel / in the design bed file) with Sufficient Depth of Base Coverage=1652887
    Average Coverage=1300.0
    Total Exonic Bases with Sufficient Depth of Base Coverage=1198355
    Denominator used for TMB Calculation (as specified in the workflow parameters)=1198355
    Total Somatic Filtered Variants Count (numerator for TMB calculation)=19
    ('missense=18,frameshiftDeletion=0, frameshiftInsertion=0, nonframeshiftDeletion=0, nonframeshiftInsertion=0, nonsense=1')
    Germline Calibration='Not Applied'
    Germline Calibration Slope=1.4637
    Germline Calibration Intercept=0.0
    TMB Standardization='Not Applied'
    TMB Standardization Slope=1.0
    TMB Standardization Intercept=0.0
    Mutation Load (Mutations/Mb)=15.86
    TMB classification (based on specified parameters)='Intermediate
    'Variant count (Germline + somatic)=1098
    Variant count: 19COSMIC Annotated Somatic Variants=2
    Variant count=19
Example: Tumor Mutational Burden calculation
Tumor Mutational Burden calculation
Ion Reporter™ Software uses TMB Algorithm v4.0:
precalibration TMB = (SM x 106)
                                   Total Exonic Bases with Sufficient Coverage
where
  • TMB is Tumor Mutational Burden
  • SM is Somatic Mutations
You can control how somatic mutations are calculated. You can also select between Exonic bases or all of the genomic bases that are covered by the panel. Similarly, the ‘Sufficient Coverage’ threshold can be modified. By default, the TMB calculation are done using Total Exonic Bases with ≥60bp coverage.
The following settings apply:
  • applyTMBStandardization: Customizable parameter (default value: off)
  • standardization intercept: TMB Standardization Factor: Intercept (default value: 0)
  • standardization slope: TMB Standardization Factor: Slope (default value: 1)
It is apparent from the values that even if the standardization is enabled with the current default values, the TMB score value are not be adjusted until a value is provided for the intercept and the slope.
Note: Standardization is independent of the germline calibration.