Create an Analysis Results table preference
You can create a table preference for the Analysis Results table in Ion Reporter™ Software. The table preference enables you to view only the columns of data that are relevant to your research.
The user-defined table preference setting is included as output when you download a variants file.
- In the Analyses tab, click Overview.
- Click the link for an analysis of interest to open the Analysis Results table.
-
Click , then select the analysis attributes that you want to add to the Analysis Results table, or deselect the analysis attributes that you want to remove from the table.
The selected columns are added on the right side of the table.
-
To save the table as a preference:
Option
Description
.
This setting enables you to name the table preference. The preference setting is then selectable from .
This setting becomes the standard view for the user account. To restore default table columns, click .
The table preference remains until you change it by clicking Restore Defaults or by applying another table preference.