Delete an Analysis Results table preference
You can delete a user-defined table preference setting for the Analysis Results table in Ion Reporter™ Software.
  1. In the Analyses tab, click Overview.
  2. Click the link for an analysis of interest to open the Analysis Results table.
  3. Click PreferencesTable Preferences, then select the user-defined table preference that you want to delete.
  4. Click PreferencesDelete Table Preferences.
  5. In the Confirm Delete dialog box, click Yes to confirm the deletion.
The user-defined table preference is removed from the Table Preferences list.