Add attribute columns to the Analysis Results table
You can add columns to the Analysis Results table that you view in Ion Reporter™ Software.
  1. In the Analyses tab, click Overview.
  2. Click the link for an analysis of interest to open the Analysis Results table.
  3. Click PreferencesSelect Columns, then select the columns that you want to add to the Analysis Results table, or deselect the columns that you want to remove from the table.
The selected columns are added to the Analysis Results table until you close the table. To save the table with the attributes that you added, see Create an Analysis Results table preference.