Change the order and width of columns in the Analysis Results table
You can change the order and width of columns in the Analysis Results table in Ion Reporter™ Software.
  1. In the Analyses tab, select Overview.
  2. Click the link for an analysis of interest to open the Analysis Results table.
  3. Click any column heading that you want to move, then drag and drop each column heading to the position that you prefer. Change the width of each column, as desired.
    The column order and width remains until you close the screen, or can optionally be saved as a table preference.
  4. (Optional) To save the order and width of the columns as a table preference:
    1. Click PreferencesSave Table Preference As.
    2. Enter a name for the table preference, then click Save.
    The Analysis Results table preference is saved.