Import MyVariants
You can import MyVariants from a VCF or a TSV file that is exported from another MyVariants database, or from a TSV file that you create. For more information, see Export MyVariants database.
- In the Analyses tab, click MyVariants, then click Import MyVariants.
- In the Upload MyVariants File screen, click Select file , then browse to the VCF or TSV file.
- Click Submit.