Classifications and Notes
Variants that use the Important or Ignore flags are often variants that can also be categorized with Classifications and include other information that can be captured in Notes. Ion Reporter™ Software includes an optional setting to add Classifications and Notes that were used in a previous occurrence of a variant automatically to subsequent occurrence of the same variant. Variants in a MyVariants database that include the setting, when reported in a new set of analysis results, automatically add the Classifications and Notes. The analysis workflow that is used for the analysis must be associated with a MyVariants database.
For example, in a MyVariants database, you can set a variant to always be flagged as Important, classified as Benign, and include an informational note.
MyVariants Flags, Classifications, and Notes are all stored in the MyVariants database. The preference to add variant Classifications and Notes automatically is disabled by default. For more information, see "Automatically add classifications and notes to variants".