View a MyVariants database associated with an analysis workflow
You can determine which MyVariants database is selected for an analysis workflow in Ion Reporter Software.
  1. In the Workflows tab, click Overview.
  2. Click the row for the analysis workflow of interest.
  3. In the Details pane, look for MyVariants Database.