Automatically add classifications and notes to variants
You can set a preference to add notes and classifications automatically to MyVariants in Ion Reporter™ Software. Use the setting to add Classifications and Notes that were used in a previous occurrence of a variant automatically to subsequent occurrence of the same variant. Variants in a MyVariants database that include the setting, when reported in a new set of analysis results, automatically add the Classifications and Notes to the variant. The analysis workflow that is used for the analysis must be associated with a MyVariants database.
- In the Analyses tab, in the MyVariants screen, click Preferences.
-
In the Preferences dialog box, select one or both options to be added to variants.
Option
Description
Notes
Information for the variant that is entered by a user.
The last note added to that same variant is added automatically to new instances of that same variant in a new analysis.
Classifications
A category that users can add to a variant.
Classification options include: Unclassified, Pathogenic, Likely Pathogenic, VUS, Uncertain Significance, Likely Benign, Benign, Unknown, Suspected Benign, Suspected Deleterious, Deleterious, Technical Artifact, Oncogenic, and Likely Oncogenic.
By default, classifications for each variant are blank until a value is selected.