Associate a MyVariants database with an analysis workflow
You can apply MyVariants that are included in a MyVariants database to the analysis results in Ion Reporter™ Software if you select a MyVariants database when you create an analysis workflow. MyVariants allows you to track variants that are important to your research and to ignore other variants that are not relevant to your research.
Each analysis workflow can be associated with only one MyVariants database in the Annotation workflow step. To check that the MyVariants database was associated with an analysis workflow, see View a MyVariants database associated with an analysis workflow.