MyVariants database
A MyVariants database is a repository of identified variants that is shared by an entire Ion Reporter Software organization, a specific research project, or an individual researcher. When you label the variants that are included in analysis results from predefined analysis workflows as Important or Ignore, the variants are automatically added to the default MyVariants database for the organization in Ion Reporter Software. This database is named MyVariantsDb_hg19. You can also create a different MyVariants database for a specific research project and then select this MyVariants database when you create an analysis workflow.
Other ways to use MyVariants databases include: