Edit a MyVariants database
You can edit a MyVariants database in Ion Reporter™ Software.
IMPORTANT! Edits to MyVariants can affect the MyVariants settings that are made by other members of the organization if you are sharing a MyVariants database.
You can change the Important or Ignore flags that are associated with variants in a MyVariants database.
If you delete an analysis that includes a variant that is labeled as Important or Ignore, the MyVariants database is unaffected and the variant flags, notes, and classifications remain.
You can also add notes to variants in a MyVariants database.
- In the Analyses tab, click MyVariants.
- In the MyVariants table, select a specific MyVariants database from the list.
-
Review the flags, notes and classifications for each variant, then update each option, if needed.