View the variants in a MyVariants database
You can view a list of variants that are contained in a specific MyVariants database in Ion Reporter™ Software. When you view variants, you can also add notes, synch the MyVariants database, generate a report, and select variants and perform batch actions.
- In the Analyses tab, click MyVariants.
- In the database list, select the MyVariants database of interest.
- View the list.