This filter can be added to filter chains in the software to report on specific genes.
Filter conditions include the ability to select all (In), none (Not In), or specific gene symbols from the list, and to include or exclude unannotated variants in filtered analysis results.
This filter returns the gene symbols of genes that are found in your samples.