Gene Ontology (GO) filter
This filter can be added to filter chains in the software to detect various gene ontologies (functions of specific genes). The specific filters are as follows.
-
GRch38 reference genome:
-
Gene Ontology (20151216)
-
Gene Ontology (20190930)
-
Gene Ontology (20200511)
-
Gene Ontology (20201118)
-
Gene Ontology (20210830)
-
-
hg19 reference genome:
-
Gene Ontology (20190930)
-
Gene Ontology (20200511)
-
Gene Ontology (20201118)
-
Gene Ontology (20210830)
-
Gene Ontology (20230314)
-
Filter conditions include the ability to select all (In), none (Not In), or to select specific annotations, and to include or exclude unannotated variants in filtered analysis results.
This filter returns gene ontologies of samples based on the selections made.