DGV filter
This filter can be added to filter chains in the software to detect human genomic structural variants that match those in the Database of Genomic Variants (DGV). The specific filters are as follows.
  • DGV filter(20130723)
  • DGV filter(20160515)
Filter conditions include the ability to include all DGV variants (In), or to exclude all DGV variants (Not In) in filtered analysis results.
The filter detects human genomic structural variants as defined by DGV, which can be included in or excluded from filtered analysis results.