COSMIC filter
This filter can be added to filter chains in the software to compare variants to the catalog of somatic mutations in tumor tissue as compiled by the COSMIC database. The specific databases are as follows.
  • COSMIC(85)
  • COSMIC(89)
Filter conditions include the ability to select all COSMIC values, to select specific annotation values, and to include or exclude unannotated variants in filtered analysis results.
The filter returns variants that match those in the COSMIC database.