Minor Allele Frequency filter
This filter can be added to filter chains in the software to detect variants with minor allele frequencies that match those in the dbSNP database from the 1000 genomes project.
Filter conditions include the ability to set a range from 0.0 to 0.5 for minor allele frequencies. In addition, the range boundary values can be included or excluded, and unannotated variants can be included or excluded in filtered analysis results.
The filter returns variants in samples that match those in the dbSNP database from the 1000 genomes project.