dbSNP filter
This filter can be added to filter chains in the software to compare single nucleotide polymorphism variants in samples against the dbSNP database. The specific filters are as follows.
  • dbSNP(151)
  • dbSNP(153)
Filter conditions include the ability to select all dbSNP values (In), or to exclude all dbSNP values (Not In).
The filter detects SNP variants that match those in the dbSNP database, which can be included in or excluded from filtered analysis results.