ExAC filters
If you use ExAC filters, it is recommended that you set the Variant View parameter to Allele View. For more information, see Locus View versus Allele View of variants.
These filters can be added to filter chains to detect rare gene variants for specific population groups in the software. The filters are derived from the Exome Aggregation Consortium (ExAC) database. The specific filters are:
-
ExAC AAF(01)
-
ExAC EAAF(01)
-
ExAC EFAF(01)
-
ExAC ENFAF(01)
-
ExAC GAF(01)
-
ExAC LAF(01)
-
ExAC OAF(01)
-
ExAC SAAF(01)
Filter conditions include the ability to set a range from 0.0 to 1.0. In addition, the range boundary values can be included or excluded, and unannotated variants can be included or excluded in filtered analysis results.
The filter detects rare gene variant results that align with the ExAC database, which can be included or excluded from filtered analysis results.