Add a filter chain to an analysis workflow
You can apply a filter chain during analysis workflow creation in Ion Reporter™ Software. Filter chains are sets of filters that you apply to variants identified in your analysis. Filters allow you to remove or include variants in your analysis results. For more information, see Filters and filter chains.
-
In the Filters step, select a default filter chain from the list.
After you select a filter chain, the filters that are contained in the filter chain are shown.
-
(Optional) Click the Workflow Presets link to configure a custom filter chain.
For more information, see Create a user-defined filter chain.
- Click Next.
The filter chain is established as the default filter chain for the analysis workflow. All future analyses that are run through this analysis workflow will use this filter chain.