Import panel files from AmpliSeq.com
You can import files for Ion AmpliSeq™ and Ion AmpliSeq™ HD Made-to-Order, On-Demand, or some Ready-to-Use, and Oncomine™ tumor specific research panels from AmpliSeq.com into Ion Reporter™ Software during analysis workflow creation. For more information, see Ion AmpliSeq and Ion AmpliSeq HD panel types.
Only ordered and quoted panel files are available for import into Ion Reporter™ Software.
- In the Research Application step, select an appropriate research application and sample group for the panel, then click Next.
-
In the Reference step, import a target regions file.
Option
Description
Custom Panel
-
Under Target Regions, click
AmpliSeq Import.
-
In the Import for AmpliSeq dialog box, select the Custom Panel tab for Ion AmpliSeq™ or Ion AmpliSeq™ HD Made-to-Order panel files, or Ion AmpliSeq™ On Demand panel files.
- Enter an account user name.
-
In the Password field, enter the access code. For more information, see Generate an access code for AmpliSeq.com.
- Click List My AmpliSeq Panels.
-
Select the panel files that you want to import from the dropdown list, then click Import.
Fixed Panel
-
Under Target Regions, click
AmpliSeq Import.
-
In the Import for AmpliSeq dialog box, select the Fixed Panel tab for Community or Ready-to-Use panel files.
-
From the list, select the panel file that you want to import.
-
Click Import.
-
-
If the analysis includes fusions, you can import fusion panel files.
Option
Description
Custom Panel
-
Under Fusion Panel, click
AmpliSeq Import.
-
In the Import for AmpliSeq dialog box, select the Custom Panel tab for Ion AmpliSeq™ or Ion AmpliSeq™ HD Made-to-Order, or Ion AmpliSeq™ On Demand fusion panel files.
-
Enter an account user name.
-
In the Password field, enter the access code. For more information, see Generate an access code for AmpliSeq.com.
-
Click List My AmpliSeq Panels.
-
From the list, select the fusion panel file that you want to import, then click Import.
Fixed Panel
-
Click
AmpliSeq Import under the Fusion Panel field.
-
In the Import for AmpliSeq dialog box, select the Fixed Panel tab for Community or Ready-to-Use panel files.
-
Enter an account user name.
-
In the Password field, enter the access code. For more information, see Generate an access code for AmpliSeq.com.
-
Click List My AmpliSeq Panels.
-
From the list, select the fusion panel file that you want to import.
-
Click Import.
-
-
Complete the remaining steps for analysis workflow creation.
For more information, see Create a new analysis workflow.
The target regions files and fusion panel files are added to the list of files that can be selected in the Reference workflow step.
Generate an access code for AmpliSeq.com
Access codes have been added to AmpliSeq.com to improve security and compatibility with single sign-on features. The access codes increase security because they are unique to each AmpliSeq.com account.
You must generate an access code and sign in with the access code each time that you import files for Ion AmpliSeq™ and Ion AmpliSeq™ HD Made-to-Order, On-Demand, or some Ready-to-Use research panels from AmpliSeq.com into Ion Reporter™ Software. The imported files are used when you create an analysis workflow for use with the AmpliSeq.com panels.
You need to generate an access code for the AmpliSeq.com account just once, if you save the access code when it is generated. If you do not save the access code, you can regenerate a new one when you sign in again on AmpliSeq.com.
You need to set a new access code when the code expires.
- Go to AmpliSeq.com, and sign in to the account.
- Click .
-
Generate an access code.