Apply an exon tile fusion baseline workflow preset to an analysis workflow
An exon tile fusion baseline is created from fusion-negative samples to establish a control baseline. For analysis workflows that include RNA Exon Tiling, you can select an exon tile fusion baseline to apply to an analysis workflow.
-
In the Workflows tab, start to create or copy an analysis workflow.
-
Click Overview, then select an analysis workflow to copy, then click .
-
Click Create.
-
-
In the Exon Tile Fusion Baseline workflow bar step, select an option from the Baseline dropdown list.
Option
Description
No Exon Tile Fusion baseline selected
If you do not want to identify RNAExon Tiles, select this option.
Exon Tile Fusion baselines
Select an existing exon tile fusion baseline from the list.
Alternatively, you can click the Workflow Presets link, then configure a new exon tile fusion baseline preset. For more information, see Create an exon tile fusion baseline workflow preset.
- Click Next.
-
Proceed through the remaining steps in the workflow bar.
For more information, see Steps in analysis workflow creation.
- In the Confirm step, click Save Workflow.