Change the default filter chain for an analysis workflow
Many Ion Reporter™ Software analysis workflows include a default filter chain. You can change the default filter chain so that your selected settings are saved with the workflow and applied automatically when the workflow is launched.
  1. Click WorkflowsCreate Workflow.
  2. In the Filters step, select a different filter chain from the list.
  3. Proceed through the remaining steps of the workflow bar, then click Save.
The newly saved filter chain is now the default filter chain for the analysis workflow. All future analyses that are run through the new version of the analysis workflow will use the new filter chain. Analyses that were previously launched with older versions of the analysis workflow are not affected when the default filter chain is changed.