Sample groups
Ion Reporter™ Software supports sample groups to use when setting up analysis workflows. The sample group selections work with research application selections, and affect the options that are available in subsequent workflow steps.
The default options are recommended. Advanced users may change these settings if needed.
|
Sample group |
Description |
Research application |
|---|---|---|
|
Paired |
Analyze and compare two samples. |
DNA |
|
Single |
Analyze a single sample. |
Aneuploidy, Annotate Variants, DNA, Fusions, DNA and Fusions, Metagenomics, Oncology-Liquid Biopsy |
|
Single Fusions |
Analyze a single fusions sample. |
Fusions |
|
Single/Multi |
Determine the microbial diversity of a 16S sample in one or more samples. |
Metagenomics |
|
Trio |
Analyze a trio of a mother, father, and proband. |
DNA |
|
Tumor—Normal |
Identify somatic mutations using advanced statistical approaches. |
DNA |