Clones detected per variable gene
Immune Repertoire analysis results are represented graphically in the Sample Results tab of the results screen for the selected analysis.
Select Clones detected per variable gene from the Views list.
The bar plot indicates the number of identified clones having a particular variable gene. Ordering of variable genes reflects position within the IGH locus.