Example time series graphs
The following Variant Label preferences were set for this example.
|
Type of chart |
Variant Label choice |
Example shown |
|---|---|---|
|
SNV/Indel |
Gene + Coding |
|
|
CNV |
Genes |
|
|
Fusions |
Genes |
|
-
Click Show All, SNV / Indel, CNV, or Fusion to view all graphs or limit the view to the selected graph.
-
Click Selected Analyses to view the list of the analyses that were selected.
-
A general key for graphs. Variant calls are represented by an open circle, hotspot reference alleles are represented by a filled square, and data points with no call are represented by a filled triangle.
-
Click Download to download a ZIP file that contains PNG files of the graph or graphs.
-
Apply Preferences to customize the graphs. For more information, see Create and view a time series graph.
-
The type of variant graph: SNV / Indel, CNV, or Fusion.
-
A key for each graph that can be customized. Click a variant label in the key to remove all data points for that variant label from the graph.
-
Hover the mouse over a data point to view the data.
-
In this example, a no call data point is shown in the second position of the SNV/Indel Time Series line graph.
-
In this example, a reference call data point is shown in the third position of the SNV/Indel Time Series line graph.