View the Mutational Signature Prediction report
To view the mutational signature prediction report (signature_identification_<analysis_name>_report.pdf) you must first download the ZIP folder. For more information, see Download filtered variants. The folder contains the following files.
-
PDF Report
-
Profile of mutational signature of sample (Available only if a mutational signature match is found.)
-
Profile of enriched COSMIC signature (One or more, available only if a mutational signature match is found.)
-
deconstructSigs Weights Pie Chart (Available only if a mutational signature match is found.)
-
Trinucleotide distribution of variants
- Extract the downloaded ZIP folder to your hard drive.
- Open the signature_prediction folder, then double-click the PDF Report file.