Reads included in Metagenomics analysis results
Note: When your metagenomics analysis workflow uses two databases, only reads that do not map to the first database are attempted with the second database. In the Results section of the screen, the DB counters column shows how many reads matched sequences in each database.
For example, in the last row, for the species vulgatus, the value 3014:26 means that 3014 reads matched sequences in the MicroSEQ™ database and 26 matched sequences in the Greengenes database.
Note: When primer information is missing in the metagenomics analysis workflow, a warning message appears during analysis review.