Locus View
Locus View is the default variant view that appears in analysis results in Ion Reporter Software, which is a locus-centric view. In Locus View, there can be multiple alternate alleles for a given locus in each line or row.