Example RNA Exon Variant graph
-
Example analysis where an RNA exon 2–7 deletion occurred in the EFGR gene. Normalized read counts for the EFGR variant that contains the intragenic fusion (EGFR-EGFR.E1E8.DelPositive.2) are higher relative to the normalized read counts for a wild type EFGR assay (EFGR.E6E7).
-
Example analysis where exon-skipping (i.e., exon deletion) of exon 14 in the MET gene was detected. Normalized read counts for the variant representative of the exon-skip event (MET‑MET.M13M15.1) are higher relative to the MET wild-type assays (for example, MET.M13M14M15.ENST00000397752.WT and MET.M17M18M19M20.ENST00000397752.WT).