Example correlation and proportion of shared clones
This is an example of a Comparison Plots tab scatterplot. The illustration indicates the frequency of clones across two samples. Frequency values are log10 transformed with a pseudocount frequency of 1E-6 added to each value. Place the pointer over a point to see the CDR3NT and AA sequence of a clone, the variable mutation, and the frequency in either sample.