Example Jaccard Similarity Index for selected samples
This is an example of a Comparison Plots tab showing the Jaccard Similarity Index for selected samples. The Jaccard similarity index is determined for each pairwise comparison and displayed in heatmap form. The Jaccard Similarity Index ranges from 0 to 1 and is calculated as the total number of shared clones divided by the total number of distinct clones across two samples.