V-gene usage and Shannon Diversity plot example
IGH V-gene usage and Shannon Diversity
Select Shannon Diversity for a view of a spectratyping plot that highlights the Shannon Diversity (entropy) of identified clones.
This is an example of an IGH spectratyping plot from an Oncomine BCR IGH‑LR Assay analysis. Circle color indicates the Shannon Diversity of clones having a particular variable gene-CDR3 nucleotide length combination. Portions of the repertoire containing highly expanded clones typically have a corresponding low Shannon Diversity value. You can further partition the clones by isotype in Ion Reporter Software with the buttons that are above the interactive spectratyping plot.