Download spectratyping overview plots
- In the Analyses tab, click Overview.
- Click the analysis name hyperlink.
-
In the Immune Repertoire Results screen, in the Actions column for the analysis of interest, click Download.
A ZIP file that contains PNG and PDF files of the plots and other results files is downloaded.