V-gene usage and evenness plot examples
IGH V-gene usage and evenness
Select Evenness for a view of a spectratyping plot that highlights evenness of identified clone sizes (Normalized Shannon Entropy).
This is an example of an immunoglobulin heavy chain (IGH) V-gene usage and evenness spectratyping plot from an Oncomine BCR IGH‑LR Assay analysis. Circle color indicates the evenness of clone sizes for clones having a particular variable gene-CDR3 nucleotide length combination. Values range from 0 to 1, with 1 indicating most even clone sizes. In this representation, portions of the repertoire containing highly expanded clones appear white. You can further partition the clones by isotype in Ion Reporter Software, with the buttons that are above the interactive spectratyping plot.
This is an example of an IGH V-gene usage and evenness spectratyping plot from an Oncomine IGHV Leader-J Assay analysis.